2-Chlor-5-formylphenylboronsäure

Lieferant: Apollo Scientific
Gefahr

Synonyme: (2-Chloro-5-formylphenyl)boronic acid

OR9492-1G OR9492-5G OR9492-25G
APOSOR9492-1GEA 21.9 EUR
APOSOR9492-1G APOSOR9492-5G APOSOR9492-25G
2-Chlor-5-formylphenylboronsäure
2-Chlor-5-formylphenylboronsäure
Formel: C₇H₆BClO₃
Molecular Weight: 184,3847 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06656269
CAS-Nummer: 1150114-78-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR