2-Chlor-5-(hydroxymethyl)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-5-hydroxymethylphenylboronic acid
OR9493-1G
OR9493-5G
OR9493-25G
APOSOR9493-1GEA
21.9
EUR
APOSOR9493-1G
APOSOR9493-5G
APOSOR9493-25G
2-Chlor-5-(hydroxymethyl)phenylboronsäure
2-Chlor-5-(hydroxymethyl)phenylboronsäure
Formel:
C₇H₈BClO₃ Molecular Weight: 186,4000 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06656270 CAS-Nummer: 1003042-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...