1-(4-Chlorophenyl)-2-methylsulphonylethanone 95+%

Lieferant: Apollo Scientific

Synonyme: 4'-Chloro-2-(methylsulphonyl)acetophenone

OR9494-1G OR9494-250MG
APOSOR9494-1GEA 39.3 EUR
APOSOR9494-1G APOSOR9494-250MG
1-(4-Chlorophenyl)-2-methylsulphonylethanone 95+%
1-(4-Chlorophenyl)-2-methylsulphonylethanone
Formel: C₉H₉ClO₃S
Molecular Weight: 232,69 g/mol
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00202209
CAS-Nummer: 24437-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR