(2-Methoxyethyl)benzol 95%
Lieferant: Apollo Scientific
Synonyme:
2-Phenylethyl methyll ether, Methyl Phenethyl ether
OR949664-25G
OR949664-500G
APOSOR949664-25GEA
132
EUR
APOSOR949664-25G
APOSOR949664-500G
(2-Methoxyethyl)benzol 95%
(2-Methoxyethyl)benzene
Formel:
(C₆H₅)CH₂CH₂OCH₃ Molecular Weight: 136,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025912 CAS-Nummer: 3558-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...