(±)-5-Methyl-2-hexanol 95%

Lieferant: Apollo Scientific

OR950043-25G OR950043-5G
APOSOR950043-25GEA 219 EUR
APOSOR950043-25G APOSOR950043-5G
(±)-5-Methyl-2-hexanol 95%
(±)-5-Methyl-2-hexanol
Formel: C₇H₁₆O
Molecular Weight: 116,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004558
CAS-Nummer: 627-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR