1-Thien-2-ylcyclopropanecarboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 1-(Thiophen-2-yl)cyclopropanecarboxylic acid

OR950102-1G OR950102-250MG
APOSOR950102-1GEA 151 EUR
APOSOR950102-1G APOSOR950102-250MG
1-Thien-2-ylcyclopropanecarboxylic acid 95%
1-Thien-2-ylcyclopropanecarboxylic acid
Formel: C₈H₈O₂S
Molecular Weight: 168,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16622241
CAS-Nummer: 162959-94-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR