2-Methyl-2-hexenoic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-Methylhex-2-enoic acid

OR950127-5G
APOSOR950127-5GEA 235 EUR
APOSOR950127-5G
2-Methyl-2-hexenoic acid 95%
2-Methyl-2-hexenoic acid
Formel: C₇H₁₂O₂
Molecular Weight: 128,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015855
CAS-Nummer: 28897-58-7
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR