N-Methyl-N-(thien-3-ylmethyl)amine 95%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-3-Thiophenemethanamine

OR950245-1G OR950245-250MG
APOSOR950245-1GEA 194 EUR
APOSOR950245-1G APOSOR950245-250MG
N-Methyl-N-(thien-3-ylmethyl)amine 95%
N-Methyl-N-(thien-3-ylmethyl)amine
Formel: C₆H₉NS
Molecular Weight: 127,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04536653
CAS-Nummer: 210552-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR