4-Nitro-D-phenylalanine

Lieferant: Apollo Scientific
Achtung

Synonyme: H-D-Phe(4-NO2)-OH, H-D-Phe(4-NO₂)-OH

OR9505-100G OR9505-25G OR9505-5G
APOSOR9505-100GEA 95.7 EUR
APOSOR9505-100G APOSOR9505-25G APOSOR9505-5G
4-Nitro-D-phenylalanine
4-Nitro-D-phenylalanine
Formel: C₉H₁₀N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00066016
CAS-Nummer: 56613-61-7
EINECS: 213-446-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR