(1-Aminocyclohexyl)acetic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-(1-Aminocyclohexyl)acetic acid

OR950533-1G OR950533-5G
APOSOR950533-1GEA 62.5 EUR
APOSOR950533-1G APOSOR950533-5G
(1-Aminocyclohexyl)acetic acid 97%
(1-Aminocyclohexyl)acetic acid
Formel: C₈H₁₅NO₂
Molecular Weight: 157,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258116
CAS-Nummer: 37631-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR