N,N-Dimethyl-2-(4-methyl-1-piperazinyl)ethanamin 95%

Lieferant: Apollo Scientific

OR950768-25G OR950768-500G
APOSOR950768-25GEA 132 EUR
APOSOR950768-25G APOSOR950768-500G
N,N-Dimethyl-2-(4-methyl-1-piperazinyl)ethanamin 95%
1-(2-Dimethylaminoethyl)-4-methylpiperazine
Formel: C₉H₂₁N₃
Molecular Weight: 171,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059773
CAS-Nummer: 104-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR