3-Amino-1-adamantanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Aminoadamantan-1-ol
OR9509-100G
OR9509-25G
OR9509-5G
OR9509-500G
APOSOR9509-100GEA
24.8
EUR
APOSOR9509-100G
APOSOR9509-25G
APOSOR9509-5G
APOSOR9509-500G
3-Amino-1-adamantanol
3-Amino-1-adamantanol
Formel:
C₁₀H₁₇NO Molecular Weight: 167,25 g/mol Schmelzpunkt: 263…269 °C (Zers.) Flammpunkt: 110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01821204 CAS-Nummer: 702-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...