Phenyl propionate 95%

Lieferant: Apollo Scientific

Synonyme: Propionic Acid Phenyl Ester

OR950919-25G OR950919-5G
APOSOR950919-25GEA 120 EUR
APOSOR950919-25G APOSOR950919-5G
Phenyl propionate 95%
Phenyl propionate
Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
Siedepunkt: 211 °C
Dichte: 1,05 g/cm³ (25 °C)
Flammpunkt: 86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053765
CAS-Nummer: 637-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR