4-Bromo-6-methylpyrimidine 96%

Lieferant: Apollo Scientific

OR950939-1G OR950939-250MG
APOSOR950939-1GEA 136 EUR
APOSOR950939-1G APOSOR950939-250MG
4-Bromo-6-methylpyrimidine 96%
4-Bromo-6-methylpyrimidine
Formel: C₅H₅BrN₂
Molecular Weight: 173.0106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234065
CAS-Nummer: 69543-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR