4-Methoxyphenylacetamide 97%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Methoxyphenyl)acetamide

OR951079-25G
APOSOR951079-25GEA 161 EUR
APOSOR951079-25G
4-Methoxyphenylacetamide 97%
4-Methoxyphenylacetamide
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017144
CAS-Nummer: 6343-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR