4-Phenyl-1,3-thiazole-2-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Phenylthiazole-2-carboxylic acid
OR951137-1G
OR951137-5G
APOSOR951137-1GEA
215
EUR
APOSOR951137-1G
APOSOR951137-5G
4-Phenyl-1,3-thiazole-2-carboxylic acid 95%
4-Phenyl-1,3-thiazole-2-carboxylic acid
Formel:
C₁₀H₇NO₂S Molecular Weight: 205.24 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00990415 CAS-Nummer: 59020-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...