trans-4-Methylcyclohexyl isocyanate 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
trans-4-methycyclohexyl isocyanate, trans-4-Methylcyclohexylisocyanate
OR951159-100G
APOSOR951159-100GEA
115
EUR
APOSOR951159-100G
trans-4-Methylcyclohexyl isocyanate 97%
trans-4-Methylcyclohexyl isocyanate
- May form small amount of sediment.
Formel:
C₈H₁₃NO Molecular Weight: 139,198 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD06411231 CAS-Nummer: 32175-00-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...