2-Chloro-1-(1-methyl-1H-pyrrol-2-yl)ethanone 95%

Lieferant: Apollo Scientific

OR951163-1G OR951163-250MG
APOSOR951163-1GEA 754 EUR
APOSOR951163-1G APOSOR951163-250MG
2-Chloro-1-(1-methyl-1H-pyrrol-2-yl)ethanone 95%
2-Chloro-1-(1-methyl-1H-pyrrol-2-yl)ethanone
Formel: C₇H₈ClNO
Molecular Weight: 157,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06357821
CAS-Nummer: 23694-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR