3-(1-Hydroxyethyl)phenol 98%
Lieferant: Apollo Scientific
Synonyme:
3-(1-Hydroxyethyl)phenol, 1-(3-Hydroxyphenyl)ethanol
OR951348-5G
APOSOR951348-5GEA
94.4
EUR
APOSOR951348-5G
3-(1-Hydroxyethyl)phenol 98%
3-(1-Hydroxyethyl)phenol
Formel:
C₈H₁₀O₂ Molecular Weight: 138,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00238621 CAS-Nummer: 2415-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...