4-Ethylthiobenzamid 95%

Lieferant: Apollo Scientific

Synonyme: 4-Ethylthiobenzoesäureamid

OR951373-1G OR951373-250MG
APOSOR951373-1GEA 435 EUR
APOSOR951373-1G APOSOR951373-250MG
4-Ethylthiobenzamid 95%
4-Ethylthiobenzamid
Formel: C₉H₁₁NS
Molecular Weight: 165,26 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 57774-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR