Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 1,2,3,4-tetrahydroquinoline-6-carboxylate

OR951387-1G OR951387-5G
APOSOR951387-1GEA 56.7 EUR
APOSOR951387-1G APOSOR951387-5G
Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylate 97%
Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylate
Formel: C₁₁H₁₃NO₂
Molecular Weight: 191,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08692534
CAS-Nummer: 177478-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR