3-(Benzylamino)-1-propanol 96%

Lieferant: Apollo Scientific

Synonyme: 3-(Benzylamino)propan-1-ol, N-(3-Hydroxypropyl)benzylamine, 3-(Benzylamino)-1-propanol

OR951436-100G OR951436-25G
APOSOR951436-100GEA 244 EUR
APOSOR951436-100G APOSOR951436-25G
3-(Benzylamino)-1-propanol 96%
3-(Benzylamino)-1-propanol
Formel: C₁₀H₁₅NO
Molecular Weight: 165,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020622
CAS-Nummer: 4720-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR