4-(Methylsulphonyl)benzyl alcohol 98%

Lieferant: Apollo Scientific

Synonyme: 4-(Methylsulphonyl)benzyl Alcohol

OR951461-25G
APOSOR951461-25GEA 71.2 EUR
APOSOR951461-25G
4-(Methylsulphonyl)benzyl alcohol 98%
4-(Methylsulphonyl)benzyl alcohol
Formel: C₈H₁₀O₃S
Molecular Weight: 186,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425695
CAS-Nummer: 22821-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR