5,6,7,8-Tetrahydroimidazo[1,2-a]pyridine-2-carboxylic acid 98%

Lieferant: Apollo Scientific

OR951470-1G OR951470-250MG
APOSOR951470-1GEA 252 EUR
APOSOR951470-1G APOSOR951470-250MG
5,6,7,8-Tetrahydroimidazo[1,2-a]pyridine-2-carboxylic acid 98%
5,6,7,8-Tetrahydroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,1772 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10566765
CAS-Nummer: 917364-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR