2-(3-Methoxyphenoxy)ethanamine 97%
Lieferant: Apollo Scientific
Synonyme:
2-(3-Methoxyphenoxy)ethylamine
OR951497-1G
OR951497-5G
APOSOR951497-1GEA
133
EUR
APOSOR951497-1G
APOSOR951497-5G
2-(3-Methoxyphenoxy)ethanamine 97%
2-(3-Methoxyphenoxy)ethanamine
Formel:
C₉H₁₃NO₂ Molecular Weight: 167,21 g/mol Siedepunkt: 152…154 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02598966 CAS-Nummer: 6487-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...