Ethyl-3-aminopicolinat 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-aminopicolinate, 3-Aminopyridine-2-carboxylic acid ethyl ester

OR951533-10G OR951533-5G
APOSOR951533-10GEA 88.6 EUR
APOSOR951533-10G APOSOR951533-5G
Ethyl-3-aminopicolinat 98%
Ethyl-3-aminopicolinat
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00661294
CAS-Nummer: 27507-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR