6-Chloroimidazo[1,2-b]pyridazine-3-carbaldehyde 95%
Lieferant: Apollo Scientific
OR951647-1G
OR951647-250MG
APOSOR951647-1GEA
364
EUR
APOSOR951647-1G
APOSOR951647-250MG
6-Chloroimidazo[1,2-b]pyridazine-3-carbaldehyde 95%
6-Chloroimidazo[1,2-b]pyridazine-3-carbaldehyde
Formel:
C₇H₄ClN₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044762 CAS-Nummer: 154578-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...