2-Ethoxy-2-methylpropanoic acid 95%

Lieferant: Apollo Scientific

OR951707-1G OR951707-250MG
APOSOR951707-1GEA 647 EUR
APOSOR951707-1G APOSOR951707-250MG
2-Ethoxy-2-methylpropanoic acid 95%
2-Ethoxy-2-methylpropanoic acid
Formel: C₆H₁₂O₃
Molecular Weight: 132.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14636712
CAS-Nummer: 15001-71-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR