2-Chloro-1-(2,3-dihydrobenzofuran-5-yl)ethanone ≥98%
Lieferant: Apollo Scientific
OR951841-1G
OR951841-5G
APOSOR951841-1GEA
415
EUR
APOSOR951841-1G
APOSOR951841-5G
2-Chloro-1-(2,3-dihydrobenzofuran-5-yl)ethanone ≥98%
2-Chloro-1-(2,3-dihydrobenzofuran-5-yl)ethanone
Formel:
C₁₀H₉ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03618417 CAS-Nummer: 64089-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...