Methyl-5-formylthiophene-2-carboxylate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-formylthiophene-2-carboxylate, 5-Formylthiophene-2-carboxylic acid methyl ester
OR951877-25G
OR951877-5G
APOSOR951877-25GEA
594
EUR
APOSOR951877-25G
APOSOR951877-5G
Methyl-5-formylthiophene-2-carboxylate 98%
Methyl-5-formylthiophene-2-carboxylate
Formel:
C₇H₆O₃S Molecular Weight: 170,1900 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01859942 CAS-Nummer: 67808-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...