2-Acetamido-5-methoxytoluene 95%
Lieferant: Apollo Scientific
Synonyme:
4'-Methoxy-2'-methylacetanilid
OR951990-1G
OR951990-25G
OR951990-5G
APOSOR951990-1GEA
109
EUR
APOSOR951990-1G
APOSOR951990-25G
APOSOR951990-5G
2-Acetamido-5-methoxytoluene 95%
2-Acetamido-5-methoxytoluene
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 179,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00458126 CAS-Nummer: 31601-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...