(2-Methoxy-4-pyridyl)methanamine 98%

Lieferant: Apollo Scientific

Synonyme: (2-Methoxypyridin-4-yl)methanamine

OR952055-1G
APOSOR952055-1GEA 116 EUR
APOSOR952055-1G
(2-Methoxy-4-pyridyl)methanamine 98%
(2-Methoxy-4-pyridyl)methanamine
Formel: C₇H₁₀N₂O
Molecular Weight: 138.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09054679
CAS-Nummer: 148900-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR