Benzylpropylamine 98%

Lieferant: Apollo Scientific

Synonyme: N-Benzylpropan-1-amine

OR952076-10G OR952076-1G OR952076-5G
APOSOR952076-10GEA 502 EUR
APOSOR952076-10G APOSOR952076-1G APOSOR952076-5G
Benzylpropylamine 98%
Benzylpropylamine
Formel: C₁₀H₁₅N
Molecular Weight: 149,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015197
CAS-Nummer: 2032-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR