4,6-Dimethyl-1H-indole-2-carboxylic acid 98%

Lieferant: Apollo Scientific

OR952086-1G OR952086-250MG
APOSOR952086-1GEA 116 EUR
APOSOR952086-1G APOSOR952086-250MG
4,6-Dimethyl-1H-indole-2-carboxylic acid 98%
4,6-Dimethyl-1H-indole-2-carboxylic acid
Formel: C₁₁H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02664431
CAS-Nummer: 383132-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR