Isopropyl-(2Z)-3-amino-2-butenoat 95%

Lieferant: Apollo Scientific

Synonyme: Isopropyl 3-Aminocrotonate, Isopropyl 3-aminobut-2-enoate, 3-Aminocrotonic acid Isopropyl ester, Isopropyl 3-Amino-2-butenoate, 3-Amino-2-butenoic acid Isopropyl ester

OR952106-500G
APOSOR952106-500GEA 125 EUR
APOSOR952106-500G
Isopropyl-(2Z)-3-amino-2-butenoat 95%
Isopropyl-3-aminobut-2-enoate
Formel: C₇H₁₃NO₂
Molecular Weight: 143,1836 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00134272
CAS-Nummer: 14205-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR