2-Chloro-5-methylnicotinonitrile 98%
Lieferant: Apollo Scientific
OR952281-1G
OR952281-250MG
APOSOR952281-1GEA
632
EUR
APOSOR952281-1G
APOSOR952281-250MG
2-Chloro-5-methylnicotinonitrile 98%
2-Chloro-5-methylnicotinonitrile
Formel:
C₇H₅ClN₂ Molecular Weight: 152.58 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375374 CAS-Nummer: 66909-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...