Methyl-2-chloro-5-methylnicotinate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-chloro-5-methylpyridine-3-carboxylate, Methyl 2-chloro-5-methylnicotinate
OR952283-1G
OR952283-250MG
APOSOR952283-1GEA
106
EUR
APOSOR952283-1G
APOSOR952283-250MG
Methyl-2-chloro-5-methylnicotinate 98%
Methyl-2-chloro-5-methylnicotinate
Formel:
C₈H₈ClNO₂ Molecular Weight: 185,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375372 CAS-Nummer: 65169-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...