3-Dimethoxymethyl-2-methoxy-pyridine 95%
Lieferant: Apollo Scientific
OR952352-1G
OR952352-250MG
APOSOR952352-1GEA
451
EUR
APOSOR952352-1G
APOSOR952352-250MG
3-Dimethoxymethyl-2-methoxy-pyridine 95%
3-Dimethoxymethyl-2-methoxypyridine
Formel:
C₉H₁₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366720 CAS-Nummer: 869735-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...