6-Methoxy-5-indanecarbaldehyde 95%

Lieferant: Apollo Scientific

OR952430-1G OR952430-5G
APOSOR952430-1GEA 168 EUR
APOSOR952430-1G APOSOR952430-5G
6-Methoxy-5-indanecarbaldehyde 95%
6-Methoxy-5-indanecarbaldehyde
Formel: C₁₁H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06248004
CAS-Nummer: 73615-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR