Pseudopelletierine 95%

Lieferant: Apollo Scientific

Synonyme: 9-Methyl-9-azabicyclo[3.3.1]nonan-3-one

OR952553-1G OR952553-5G
APOSOR952553-1GEA 53.8 EUR
APOSOR952553-1G APOSOR952553-5G
Pseudopelletierine 95%
Pseudopelletierine
Formel: C₉H₁₅NO
Molecular Weight: 153,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078161
CAS-Nummer: 552-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR