Methyl-1-aminocyclopropane-1-carboxylate 96%

Lieferant: Apollo Scientific

Synonyme: Methyl 1-aminocyclopropanecarboxylate

OR952598-5G
APOSOR952598-5GEA 84.1 EUR
APOSOR952598-5G
Methyl-1-aminocyclopropane-1-carboxylate 96%
Methyl-1-aminocyclopropane-1-carboxylate
Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00906963
CAS-Nummer: 72784-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR