Ethyl-2-morpholinoacetate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-morpholinoacetate, Ethyl morpholinoacetate

OR952661-25G
APOSOR952661-25GEA 95.7 EUR
APOSOR952661-25G
Ethyl-2-morpholinoacetate 95%
Ethyl-2-morpholinoacetate
Formel: C₈H₁₅NO₃
Molecular Weight: 173,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023375
CAS-Nummer: 3235-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR