Methyl-5-Methoxy-3-Oxovalerat 95%
Lieferant: Apollo Scientific
Synonyme:
5-Methoxy-3-oxovaleric Acid Methyl Ester, Methyl 3-Keto-5-methoxyvalerate, 3-Keto-5-methoxyvaleric Acid Methyl Ester, Methyl 5-methoxy-3-oxopentanoate
OR952690-1G
OR952690-5G
APOSOR952690-1GEA
92.8
EUR
APOSOR952690-1G
APOSOR952690-5G
Methyl-5-Methoxy-3-Oxovalerat 95%
Methyl-5-Methoxy-3-Oxovalerat
Formel:
C₇H₁₂O₄ Molecular Weight: 160,17 g/mol Siedepunkt: 56…57 °C/0,075mmHg Dichte: 1,1 g/cm³ (25 °C) Flammpunkt: 104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042840 CAS-Nummer: 62462-05-9 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...