1-Ethylbenzotriazol-5-carbonsäure 98%

Lieferant: Apollo Scientific

Synonyme: 1-Ethyl-1H-1,2,3-benzotriazole-5-carboxylic acid

OR952731-1G OR952731-250MG
APOSOR952731-1GEA 364 EUR
APOSOR952731-1G APOSOR952731-250MG
1-Ethylbenzotriazol-5-carbonsäure 98%
1-Ethylbenzotriazole-5-carboxylic acid
Formel: C₉H₉N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05270868
CAS-Nummer: 691363-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR