6-Chloro-1,3-benzoxazol-2-amine 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-6-chlorobenzoxazole

OR952811-5G
APOSOR952811-5GEA 95.7 EUR
APOSOR952811-5G
6-Chloro-1,3-benzoxazol-2-amine 97%
6-Chloro-1,3-benzoxazol-2-amine
Formel: C₇H₅ClN₂O
Molecular Weight: 168.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01664218
CAS-Nummer: 52112-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR