3',5'-Dihydroxyacetophenon 98%
Lieferant: Apollo Scientific
Synonyme:
1-(3,5-Dihydroxyphenyl)ethan-1-on
OR952849-100G
OR952849-500G
APOSOR952849-100GEA
91.5
EUR
APOSOR952849-100G
APOSOR952849-500G
3',5'-Dihydroxyacetophenon 98%
3',5'-Dihydroxyacetophenon
Formel:
C₈H₈O₃ Molecular Weight: 152,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002290 CAS-Nummer: 51863-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...