Ethyl-2,3-dimethylbenzoat 95%

Lieferant: Apollo Scientific

OR952926-1G OR952926-25G OR952926-5G
APOSOR952926-1GEA 102 EUR
APOSOR952926-1G APOSOR952926-25G APOSOR952926-5G
Ethyl-2,3-dimethylbenzoat 95%
Ethyl-2,3-dimethylbenzoat
Formel: C₁₁H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00017523
CAS-Nummer: 104175-24-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR