5-Methoxyisoxazol-3-amine 97%
Lieferant: Apollo Scientific
Synonyme:
3-Amino-5-methoxyisoxazole
OR952969-1G
OR952969-250MG
OR952969-5G
APOSOR952969-1GEA
723
EUR
APOSOR952969-1G
APOSOR952969-250MG
APOSOR952969-5G
5-Methoxyisoxazol-3-amine 97%
5-Methoxyisoxazol-3-amine
Formel:
C4H6N2O2 Lagertemperatur: Raumtemperatur |
CAS-Nummer:
32326-25-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...