2-Phenylisobuttersäure 98%
Lieferant: Apollo Scientific
Synonyme:
2-Methyl-2-phenylpropansäure
OR952994-500G
APOSOR952994-500GEA
102
EUR
APOSOR952994-500G
2-Phenylisobuttersäure 98%
2-Phenylisobuttersäure
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014332 CAS-Nummer: 826-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...