4-Ethoxy-3-nitropyridin 97%

Lieferant: Apollo Scientific

OR9530-5G
APOSOR9530-5GEA 58 EUR
APOSOR9530-5G
4-Ethoxy-3-nitropyridin 97%
4-Ethoxy-3-nitropyridin
Formel: C₇H₈N₂O₃
Molecular Weight: 168,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234959
CAS-Nummer: 1796-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR